Gene Transcripts

Gene Symbol
LINC01315
Entrez Gene ID
102723775
Gene Name
long intergenic non-protein coding RNA 1315
Gene Aliases
lnc-C22orf32-1
Chromosome Location
Chr. 22: 42364400 - 42369208 on Build GRCh38
UniGene ID
Hs.335413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_120595.1 2 1113
NR_120596.1 2 694
NR_120597.1 2 351

View Full Product Details