Gene Transcripts

Gene Symbol
PCDHB3
Entrez Gene ID
56132
Gene Name
protocadherin beta 3
Gene Aliases
PCDH-BETA3
Chromosome Location
Chr. 5: 141100242 - 141103827 on Build GRCh38
UniGene ID
Hs.508987
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018937.4 NP_061760.1 1 624
GenBank mRNA AF152496.1 - 1 216
AF217755.1 - 1 216
AK313451.1 - 1 393
BC104817.1 - 1 243 8143820

View Full Product Details