Gene Transcripts
- Gene Symbol
- NBPF6
- Entrez Gene ID
- 653149
- Gene Name
- neuroblastoma breakpoint family member 6
- Gene Aliases
- Chromosome Location
- Chr. 1: 108421496 - 108471903 on Build GRCh38
- UniGene ID
- Hs.712226
- Species
- Homo sapiens
- Gene Symbol
- LOC400768
- Entrez Gene ID
- 400768
- Gene Name
- uncharacterized LOC400768
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.12422
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC051808.2 |
- |
2 |
1023 |
|
View Full Product Details