Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020196.2 | NP_064581.2 | 2 | 109 | |
GenBank mRNA | AB026111.1 | - | 2 | 98 | |
AF226051.1 | - | 2 | 109 | ||
AF258567.1 | - | 2 | 104 | ||
AF272147.1 | - | 2 | 119 | ||
BC007208.1 | - | 2 | 91 | 2823235 |