Gene Transcripts

Gene Symbol
Coq4
Entrez Gene ID
366013
Gene Name
coenzyme Q4
Gene Aliases
Chromosome Location
Chr. 3: 8349386 - 8357719 on Build Rnor_6.0
UniGene ID
Rn.162647
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031662.1 NP_001026832.1 5, 6 536
GenBank mRNA BC099123.1 - 5, 6 536

View Full Product Details