Gene Transcripts

Gene Symbol
St8sia1
Entrez Gene ID
20449
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1
Gene Aliases
9330109E03Rik, GD3S, Sia-T, Siat8, Siat8a
Chromosome Location
Chr. 6: 142821541 - 142989530 on Build GRCm38
UniGene ID
Mm.260838
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011374.2 NP_035504.2 5 2974
GenBank mRNA AK046188.1 - 5 2914

View Full Product Details