Gene Transcripts

Gene Symbol
Upf3b
Entrez Gene ID
68134
Gene Name
UPF3 regulator of nonsense transcripts homolog B (yeast)
Gene Aliases
5730594O13Rik, AI317193, AW541158, RENT3B, UPF3X
Chromosome Location
Chr. X: 37091678 - 37110396 on Build GRCm38
UniGene ID
Mm.271160
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026573.2 NP_080849.1 10 1825
GenBank mRNA AK145431.1 - 10 1828
BC047049.1 - 8 1484
BC117769.1 - 5 1171 40091025
BC117770.1 - 5 1171 40091027

View Full Product Details