Gene Transcripts

Gene Symbol
MRC1
Entrez Gene ID
4360
Gene Name
mannose receptor, C type 1
Gene Aliases
CD206, CLEC13D, CLEC13DL, MMR, MRC1L1, bA541I19.1, hMR
Chromosome Location
Chr. 10: 17809343 - 17911162 on Build GRCh38
UniGene ID
Hs.743903
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002438.3 NP_002429.1 7 1339
GenBank mRNA J05550.1 - 7 1319
X55635.1 - 7 1292

View Full Product Details