Gene Transcripts

Gene Symbol
CITED4
Entrez Gene ID
163732
Gene Name
Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4
Gene Aliases
Chromosome Location
Chr. 1: 40861056 - 40862346 on Build GRCh38
UniGene ID
Hs.355820
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133467.2 NP_597724.1 1 1042
GenBank mRNA BC035496.2 - 1 1044
BC052559.1 - 1 1042 6250908

View Full Product Details