Gene Transcripts

Gene Symbol
OR5M10
Entrez Gene ID
390167
Gene Name
olfactory receptor family 5 subfamily M member 10
Gene Aliases
OR11-207
Chromosome Location
Chr. 11: 56576774 - 56577721 on Build GRCh38
UniGene ID
Hs.553750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004741.1 NP_001004741.1 1 422
GenBank mRNA BC140733.1 - 1 488

View Full Product Details