Gene Transcripts
- Gene Symbol
- Cyp2c37
- Entrez Gene ID
- 13096
- Gene Name
- cytochrome P450, family 2. subfamily c, polypeptide 37
- Gene Aliases
- Chromosome Location
- Chr. 19: 39992424 - 40012243 on Build GRCm38
- UniGene ID
- Mm.38963
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF047542.1 |
- |
|
1756 |
|
BC057912.1 |
- |
|
1757 |
5052873 |
View Full Product Details