Gene Transcripts

Gene Symbol
CXCR3
Entrez Gene ID
2833
Gene Name
C-X-C motif chemokine receptor 3
Gene Aliases
CD182, CD183, CKR-L2, CMKAR3, GPR9, IP10-R, Mig-R, MigR
Chromosome Location
Chr. X: 71615913 - 71618517 on Build GRCh38
UniGene ID
Hs.198252
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142797.1 NP_001136269.1 2 757
NM_001504.1 NP_001495.1 2 513
XM_005262256.3 XP_005262313.1 2 614
XM_005262257.3 XP_005262314.1 2 614
XM_017029435.1 XP_016884924.1 2 695
XM_017029436.1 XP_016884925.1 2 511
GenBank mRNA AF469635.1 - 2 750
AK313679.1 - 2 507
AY242128.1 - 1 445
BC034403.1 - 2 483 5176136
KU178101.1 - 1 445
KU178102.1 - 1 445
X95876.1 - 2 513

View Full Product Details