Gene Transcripts

Gene Symbol
C2orf54
Entrez Gene ID
79919
Gene Name
chromosome 2 open reading frame 54
Gene Aliases
Chromosome Location
Chr. 2: 240886048 - 240896889 on Build GRCh38
UniGene ID
Hs.193745
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085437.2 NP_001078906.2 5 1424
NM_001282921.1 NP_001269850.1 5 917
NM_024861.3 NP_079137.3 5 860
XM_011511877.1 XP_011510179.1 6 1416
GenBank mRNA AK026324.1 - 5 857
AK056601.1 - 5 1424
AK096434.1 - 5 917
BC125154.1 - 5 848 40120634
BC125155.1 - 5 848 40120635

View Full Product Details