Gene Transcripts

Gene Symbol
WDR34
Entrez Gene ID
89891
Gene Name
WD repeat domain 34
Gene Aliases
DIC5, FAP133, SRTD11, bA216B9.3
Chromosome Location
Chr. 9: 128633653 - 128656850 on Build GRCh38
UniGene ID
Hs.495240
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052844.3 NP_443076.2 9 1572
XM_011519179.2 XP_011517481.1 9 1422
GenBank mRNA BC001614.1 - 9 1407
BC011874.2 - 9 1509 3161070

View Full Product Details