Gene Transcripts
- Gene Symbol
- GOLGA3
- Entrez Gene ID
- 2802
- Gene Name
- golgin A3
- Gene Aliases
- GCP170, MEA-2
- Chromosome Location
- Chr. 12: 132768909 - 132829131 on Build GRCh38
- UniGene ID
- Hs.507333
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001172557.1 |
NP_001166028.1
|
2, 3 |
356 |
|
NM_005895.3 |
NP_005886.2
|
2, 3 |
694 |
|
XM_005266164.4 |
XP_005266221.1
|
2, 3 |
421 |
|
XM_006719736.3 |
XP_006719799.1
|
2, 3 |
421 |
|
XM_006719737.3 |
XP_006719800.1
|
2, 3 |
431 |
|
GenBank mRNA |
AB027133.1 |
- |
2, 3 |
416 |
|
AF485338.1 |
- |
1, 2 |
138 |
|
BC060826.1 |
- |
2, 3 |
380 |
|
BC142658.1 |
- |
2, 3 |
356 |
40148073 |
BC146675.1 |
- |
2, 3 |
356 |
40148852 |
D63997.1 |
- |
2, 3 |
404 |
|
View Full Product Details