Gene Transcripts

Gene Symbol
TSLP
Entrez Gene ID
85480
Gene Name
thymic stromal lymphopoietin
Gene Aliases
Chromosome Location
Chr. 5: 111070080 - 111078024 on Build GRCh38
UniGene ID
Hs.389874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033035.4 NP_149024.1 4 596
NM_138551.4 NP_612561.2 2 378
NR_045089.1 5 1801
XM_011543698.1 XP_011542000.1 3 200
GenBank mRNA AF338732.1 - 4 430
AL833421.1 - 2 358
AY037115.1 - 4 596
BC016720.1 - 2 311
BC022894.1 - 2 350
BC040592.1 - 5 1802 5269764
BF245025.1 - 2 366
BU571002.1 - 2 381

View Full Product Details