Gene Transcripts

Gene Symbol
RAB1B
Entrez Gene ID
81876
Gene Name
RAB1B, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 11: 66268526 - 66277492 on Build GRCh38
UniGene ID
Hs.300816
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030981.2 NP_112243.1 3 206
XM_017018378.1 XP_016873867.1 3 265
GenBank mRNA AF092437.1 - 3 124
AJ245875.1 - 2 111
AK292086.1 - 3 174
AK315333.1 - 3 167
AL136635.1 - 3 158
BC028331.1 - 3 157
BC071169.1 - 3 145 6026121
BG717501.1 - 3 211

View Full Product Details