Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017074.1 | NP_058770.1 | 9 | 993 | |
XM_006233536.2 | XP_006233598.1 | 8 | 932 | ||
GenBank mRNA | AB052882.2 | - | 9 | 992 | |
AY032875.1 | - | 9 | 992 | ||
AY641456.1 | - | 8 | 861 | ||
BC078869.1 | - | 9 | 1006 | 7109746 | |
D17370.1 | - | 9 | 993 | ||
X53460.1 | - | 881 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008761574.1 | XP_008759796.1 | 1040 |