Gene Transcripts

Gene Symbol
Slitrk2
Entrez Gene ID
245450
Gene Name
SLIT and NTRK-like family, member 2
Gene Aliases
A930040J07
Chromosome Location
Chr. X: 66649318 - 66661402 on Build GRCm38
UniGene ID
Mm.336081
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161431.1 NP_001154903.1 3 5278
NM_198863.2 NP_942563.2 2 5254
GenBank mRNA AK034485.1 - 1 1157
AK141924.1 - 1 2014

View Full Product Details