Gene Transcripts

Gene Symbol
Lrrc36
Entrez Gene ID
270091
Gene Name
leucine rich repeat containing 36
Gene Aliases
BC036564
Chromosome Location
Chr. 8: 105411389 - 105464086 on Build GRCm38
UniGene ID
Mm.212709
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170788.1 NP_001164259.1 3 309
GenBank mRNA BY084860.1 - 3 310

View Full Product Details