Gene Transcripts

Gene Symbol
Rftn2
Entrez Gene ID
74013
Gene Name
raftlin family member 2
Gene Aliases
2700010E02Rik, 3222401M22Rik
Chromosome Location
Chr. 1: 55170159 - 55227065 on Build GRCm38
UniGene ID
Mm.292050
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028713.1 NP_082989.1 6, 7 1580
GenBank mRNA AK012229.2 - 5, 6 1353
AK014318.1 - 6, 7 1332
BC038341.1 - 6, 7 1580 4218473

View Full Product Details