Gene Transcripts

Gene Symbol
Hnrnpa0
Entrez Gene ID
77134
Gene Name
heterogeneous nuclear ribonucleoprotein A0
Gene Aliases
1110055B05Rik, 3010025E17Rik, Hnrpa0, hnRNP A0
Chromosome Location
Chr. 13: 58125879 - 58128556 on Build GRCm38
UniGene ID
Mm.490274
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029872.1 NP_084148.1 1 1381
GenBank mRNA AK019388.1 - 1 1406
BC023150.1 - 1 213

View Full Product Details