Gene Transcripts

Gene Symbol
Hspb3
Entrez Gene ID
78951
Gene Name
heat shock protein B3
Gene Aliases
Chromosome Location
Chr. 2: 45517788 - 45518502 on Build Rnor_6.0
UniGene ID
Rn.20155
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031750.1 NP_113938.1 1 359
GenBank mRNA AF203374.1 - 1 359

View Full Product Details