Gene Transcripts

Gene Symbol
C3ar1
Entrez Gene ID
84007
Gene Name
complement component 3a receptor 1
Gene Aliases
Chromosome Location
Chr. 4: 155681767 - 155691114 on Build Rnor_6.0
UniGene ID
Rn.226511
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032060.1 NP_114449.1 2 793
XM_006237314.2 XP_006237376.1 2 1030
GenBank mRNA U86379.1 - 2 793

View Full Product Details