Gene Transcripts
- Gene Symbol
- Erp29
- Entrez Gene ID
- 117030
- Gene Name
- endoplasmic reticulum protein 29
- Gene Aliases
- Chromosome Location
- Chr. 12: 40619377 - 40625884 on Build Rnor_6.0
- UniGene ID
- Rn.32904
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053961.2 |
NP_446413.1
|
1, 2 |
167 |
|
GenBank mRNA |
BC091129.1 |
- |
1, 2 |
167 |
|
FQ211664.1 |
- |
1, 2 |
207 |
|
FQ213671.1 |
- |
1, 2 |
165 |
|
FQ218939.1 |
- |
1, 2 |
165 |
|
FQ220022.1 |
- |
1, 2 |
165 |
|
FQ233694.1 |
- |
1, 2 |
165 |
|
FQ234786.1 |
- |
1, 2 |
165 |
|
U36482.1 |
- |
1, 2 |
195 |
|
Y10264.1 |
- |
1, 2 |
166 |
|
View Full Product Details