Gene Transcripts

Gene Symbol
Ddx46
Entrez Gene ID
245957
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 46
Gene Aliases
Chromosome Location
Chr. 17: 9475275 - 9519544 on Build Rnor_6.0
UniGene ID
Rn.3436
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139098.1 NP_620798.1 2 324
XM_006253601.2 XP_006253663.1 2 354
GenBank mRNA BC107590.1 - 2 190
U25746.1 - 2 324

View Full Product Details