Gene Transcripts

Gene Symbol
Clrn1
Entrez Gene ID
261738
Gene Name
clarin 1
Gene Aliases
Ush3, Ush3a
Chromosome Location
Chr. 2: 149049925 - 149088787 on Build Rnor_6.0
UniGene ID
Rn.57072
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153299.2 NP_695211.2 3 572
GenBank mRNA AF482698.1 - 3 572

View Full Product Details