Gene Transcripts
- Gene Symbol
- Stat1
- Entrez Gene ID
- 20846
- Gene Name
- signal transducer and activator of transcription 1
- Gene Aliases
- 2010005J02Rik, AA408197
- Chromosome Location
- Chr. 1: 52119438 - 52161865 on Build GRCm38
- UniGene ID
- Mm.277406
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001205313.1 |
NP_001192242.1
|
4, 5 |
580 |
|
NM_001205314.1 |
NP_001192243.1
|
4, 5 |
616 |
|
NM_009283.4 |
NP_033309.3
|
4, 5 |
631 |
|
GenBank mRNA |
AK018544.1 |
- |
4, 5 |
582 |
|
AK039458.1 |
- |
4, 5 |
632 |
|
AK046517.1 |
- |
4, 5 |
580 |
|
AK082706.1 |
- |
4, 5 |
613 |
|
AK084855.1 |
- |
4, 5 |
565 |
|
AK088874.1 |
- |
4, 5 |
500 |
|
AK157263.1 |
- |
4, 5 |
480 |
|
AK159887.1 |
- |
4, 5 |
615 |
|
BC004808.1 |
- |
4, 5 |
567 |
3587831 |
U06924.1 |
- |
2, 3 |
285 |
|
View Full Product Details