Gene Transcripts

Gene Symbol
Zfp40
Entrez Gene ID
22700
Gene Name
zinc finger protein 40
Gene Aliases
CTfin12, NTfin12, Zfp-40
Chromosome Location
Chr. 17: 23173869 - 23193228 on Build GRCm38
UniGene ID
Mm.486936
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009555.2 NP_033581.2 6 848
GenBank mRNA AK049894.1 - 6 854
BC061205.1 - 6 817 30248495
BC065097.1 - 6 841
D10632.1 - 2 530

View Full Product Details