Gene Transcripts

Gene Symbol
Zfp53
Entrez Gene ID
24132
Gene Name
zinc finger protein 53
Gene Aliases
D030067O06Rik, KRAZ1, Zfp-53, Zfp118, zfas8
Chromosome Location
Chr. 17: 21488988 - 21510477 on Build GRCm38
UniGene ID
Mm.42140
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013843.3 NP_038871.1 5 658
GenBank mRNA AB024004.1 - 466
AK030965.1 - 5 648
AK148857.1 - 5 661
BC020362.1 - 5 564
BC046328.1 - 5 661 6475651

View Full Product Details