Gene Transcripts
- Gene Symbol
- Lpin3
- Entrez Gene ID
- 64899
- Gene Name
- lipin 3
- Gene Aliases
- 9130206L11Rik, AA438110, AV236139, mKIAA4023
- Chromosome Location
- Chr. 2: 160880670 - 160906000 on Build GRCm38
- UniGene ID
- Mm.292111
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199118.1 |
NP_001186047.1
|
2, 3 |
448 |
|
NM_022883.3 |
NP_075021.1
|
2, 3 |
525 |
|
GenBank mRNA |
AF286724.1 |
- |
2, 3 |
462 |
|
AK033647.1 |
- |
2, 3 |
446 |
|
AK049363.1 |
- |
2, 3 |
441 |
|
AK163833.1 |
- |
2, 3 |
440 |
|
AK220228.1 |
- |
2, 3 |
525 |
|
BB659464.1 |
- |
2, 3 |
525 |
|
BC026729.1 |
- |
2, 3 |
432 |
|
BC117882.1 |
- |
1, 2 |
389 |
40096038 |
BC117883.1 |
- |
1, 2 |
389 |
40096041 |
View Full Product Details