Gene Transcripts

Gene Symbol
Ceacam12
Entrez Gene ID
67315
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 12
Gene Aliases
1600031J20Rik, Ceacam12-C1, Ceacam12-C3, Ceacam12C1, Ceacam12C3
Chromosome Location
Chr. 7: 18065929 - 18077986 on Build GRCm38
UniGene ID
Mm.46089
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026087.3 NP_080363.2 4 970
GenBank mRNA AK005400.1 - 4 972
AK145926.1 - 4 921

View Full Product Details