Gene Transcripts

Gene Symbol
Rabepk
Entrez Gene ID
227746
Gene Name
Rab9 effector protein with kelch motifs
Gene Aliases
8430412M01Rik, 9530020D24Rik, AV073337, C87311, Rab9p40
Chromosome Location
Chr. 2: 34777892 - 34799912 on Build GRCm38
UniGene ID
Mm.158705
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145522.4 NP_663497.2 8 1259
GenBank mRNA AK035343.1 - 8 1139
BC019800.1 - 8 1135 5150243

View Full Product Details