Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282055.1 | NP_001268984.1 | 10 | 1501 | |
GenBank mRNA | AK088950.1 | - | 10 | 1473 | |
AK129220.1 | - | 10 | 1474 | ||
BC026492.1 | - | 3 | 446 | 4505095 | |
BC092240.1 | - | 9 | 1412 | 30662573 |