Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252520.1 | NP_001239449.1 | 21 | 3109 | |
NM_001252521.1 | NP_001239450.1 | 20 | 3100 | ||
NM_001252522.1 | NP_001239451.1 | 20 | 2974 | ||
NM_001252523.1 | NP_001239452.1 | 20 | 2959 | ||
NM_001252524.1 | NP_001239453.1 | 19 | 2935 | ||
NM_146194.4 | NP_666306.2 | 21 | 3124 | ||
GenBank mRNA | AK085472.2 | - | 16 | 2169 | |
AK144960.1 | - | 21 | 2888 | ||
AK162360.1 | - | 21 | 2802 | ||
AK165066.1 | - | 15 | 2071 | ||
AK170373.1 | - | 19 | 2955 | ||
AK220291.1 | - | 21 | 2825 | ||
BC011470.1 | - | 21 | 2819 | 2651109 | |
BC021491.1 | - | 19 | 2884 | 5345558 | |
BC023843.1 | - | 20 | 2848 | 5346041 | |
BC025566.1 | - | 20 | 2997 | 5322606 | |
BC027116.1 | - | 17 | 2396 |