Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172806.2 | NP_766394.2 | 4 | 564 | |
GenBank mRNA | AK033160.1 | - | 3 | 477 | |
AK122522.1 | - | 2 | 214 | ||
AK134271.1 | - | 3 | 415 | ||
BC041669.1 | - | 4 | 564 | 3482657 | |
BC057303.1 | - | 3 | 283 | 5702276 |