Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163713.1 | NP_001157185.1 | 3 | 354 | |
NM_172745.3 | NP_766333.1 | 3 | 354 | ||
GenBank mRNA | AK075857.1 | - | 3 | 278 | |
AK084724.1 | - | 3 | 335 | ||
AK152858.1 | - | 3 | 349 | ||
AK153135.1 | - | 3 | 349 | ||
BC060959.1 | - | 3 | 331 | 30379757 | |
BC100596.1 | - | 3 | 347 | 6762419 | |
BY246546.1 | - | 3 | 354 |