Gene Transcripts

Gene Symbol
Pla2g3
Entrez Gene ID
237625
Gene Name
phospholipase A2, group III
Gene Aliases
Chromosome Location
Chr. 11: 3487596 - 3499346 on Build GRCm38
UniGene ID
Mm.100476
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172791.2 NP_766379.2 1 328
GenBank mRNA AK033572.1 - 1 272
AK081075.1 - 1 256
BC079556.1 - 1 328 30360278

View Full Product Details