Gene Transcripts

Gene Symbol
Frmd6
Entrez Gene ID
319710
Gene Name
FERM domain containing 6
Gene Aliases
2610019M19Rik, 4930488L10Rik, AW212977
Chromosome Location
Chr. 12: 70825335 - 70902234 on Build GRCm38
UniGene ID
Mm.2962
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028127.3 NP_082403.2 3 391
GenBank mRNA AK029709.1 - 2 174
AK054457.1 - 3 776
BC053929.1 - 3 391 6389695

View Full Product Details