Gene Transcripts

Gene Symbol
Ano9
Entrez Gene ID
71345
Gene Name
anoctamin 9
Gene Aliases
5430425C04Rik, Tmem16j, Tp53i5, Trp53i5
Chromosome Location
Chr. 7: 141101218 - 141117806 on Build GRCm38
UniGene ID
Mm.235813
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178381.3 NP_848468.2 23 2279
GenBank mRNA AK017334.1 - 4 329
BC025477.1 - 22 2355
BC049815.1 - 12 2385
BY717147.1 - 4 325

View Full Product Details