Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WAKL4
Entrez Gene ID
838184
Gene Name
wall associated kinase-like 4
Gene Aliases
AT1G16150, T24D18.23, T24D18_23, wall associated kinase-like 4
Chromosome Location
Chr.1: 5532298 - 5535060 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332211.1 NP_001319018.1 1 - 2 1045 82

View Full Product Details