Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP72C1
Entrez Gene ID
838276
Gene Name
cytochrome p450 72c1
Gene Aliases
AT1G17060, CHI2, CHIBI 2, F20D23.24, F20D23_24, SHK1, SHRINK 1, SOB7, SUPPRESSOR OF PHYB-4 7, cytochrome p450 72c1
Chromosome Location
Chr.1: 5832094 - 5835488 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332275.1 NP_001319024.1 1 - 2 516 99

View Full Product Details