Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY61
Entrez Gene ID
838467
Gene Name
WRKY DNA-binding protein 61
Gene Aliases
AT1G18860, ATWRKY61, F6A14.5, F6A14_5, WRKY DNA-BINDING PROTEIN 61, WRKY DNA-binding protein 61
Chromosome Location
Chr.1: 6508797 - 6511397 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332376.1 NP_001319038.1 2 - 3 825 68

View Full Product Details