Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRCA1
Entrez Gene ID
827854
Gene Name
breast cancer susceptibility1
Gene Aliases
ARABIDOPSIS THALIANA BREAST CANCER SUSCEPTIBILITY1, AT4G21070, ATBRCA1, breast cancer susceptibility1
Chromosome Location
Chr.4: 11247991 - 11252757 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118225.5 NP_193839.4 1 - 2 235 97
GenBank mRNA AF515728.1 - 1 - 2 102 97
AY081328.1 - 1 - 2 148 97
BT010547.1 - 1 - 2 52 97

View Full Product Details