Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCFS4
Entrez Gene ID
825827
Gene Name
PCF11P-similar protein 4
Gene Aliases
AT4G04885, PCF11P-similar protein 4
Chromosome Location
Chr.4: 2471866 - 2476025 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148232.3 NP_680598.2 1 - 2 548 117
GenBank mRNA AK229122.1 - 1 - 2 442 117

View Full Product Details