Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRX
Entrez Gene ID
840078
Gene Name
DZC (Disease resistance/zinc finger/chromosome condensation-like region) domain containing protein
Gene Aliases
AT1G31880, BREVIS RADIX, F5M6.11, F5M6_11, NIP3;1, NLM9, NOD26-like intrinsic protein 3;1
Chromosome Location
Chr.1: 11445490 - 11450232 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332985.1 NP_001323364.1 2 - 3 183 74
NM_102925.3 NP_174471.2 3 - 4 510 74
GenBank mRNA BT002854.1 - 3 - 4 510 74
BT004390.1 - 1 - 2 107 74

View Full Product Details