Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HSFA1D
- Entrez Gene ID
-
840125
- Gene Name
- heat shock transcription factor A1D
- Gene Aliases
-
AT1G32330, ATHSFA1D, F27G20.6, heat shock transcription factor A1D
- Chromosome Location
-
Chr.1: 11656964 - 11660471 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102966.4 |
NP_174511.2 |
2 - 3 |
1681 |
87 |
View Full Product Details