Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MYC2
- Entrez Gene ID
-
840158
- Gene Name
- Basic helix-loop-helix (bHLH) DNA-binding family protein
- Gene Aliases
-
AT1G32640, ATMYC2, F6N18.4, F6N18_4, JAI1, JASMONATE INSENSITIVE 1, JIN1, RD22BP1, ZBF1
- Chromosome Location
-
Chr.1: 11798119 - 11801407 on Build TAIR10.1
- Gene Symbol
- AT1G32630
- Entrez Gene ID
-
840157
- Gene Name
- FAM50A-like protein
- Gene Aliases
-
AT1G32630, F6N18.3, F6N18_3
- Chromosome Location
-
Chr.1: 11797330 - 11801359 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102997.4 |
NP_174540.1 |
2 - 2 |
1196 |
158 |
View Full Product Details