Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HK1
Entrez Gene ID
816291
Gene Name
histidine kinase 1
Gene Aliases
AHK1, AT2G17820, ATHK1, T13L16.16, T13L16_16, histidine kinase 1
Chromosome Location
Chr.2: 7742870 - 7748502 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127335.2 NP_565424.1 7 - 8 2384 96

View Full Product Details