Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDL1
Entrez Gene ID
819621
Gene Name
WVD2-like 1
Gene Aliases
AT3G04630, F7O18.11, F7O18_11, WVD2-like 1
Chromosome Location
Chr.3: 1258519 - 1260871 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337538.1 NP_001319470.1 2 - 3 730 67
NM_111335.2 NP_566233.1 3 - 4 551 67
NM_180183.3 NP_850514.1 3 - 4 575 67
GenBank mRNA AK117325.1 - 3 - 4 416 67
BT024507.1 - 2 - 3 329 67

View Full Product Details